Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026364.1 | NP_080640.1 | 1, 2 | 301 | ||
GenBank mRNA | AK011290.1 | - | 1, 2 | 271 | ||
AK077439.1 | - | 1, 2 | 304 | |||
AK082484.1 | - | 1, 2 | 270 | |||
AK160763.1 | - | 1, 2 | 285 | |||
AK218181.1 | - | 1, 2 | 35 | |||
BC029621.1 | - | 1, 2 | 292 | 4208743 | ||
BY261597.1 | - | 1, 2 | 303 |