Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146342.2 | NP_001139814.2 | 3 | 534 | ||
NM_001167818.1 | NP_001161290.1 | 5 | 783 | |||
GenBank mRNA | AK170321.1 | - | 3 | 535 | ||
BC052355.1 | - | 5 | 614 | 6770333 | ||
DQ788834.1 | - | 5 | 560 |