Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pllp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026385.4 | NP_080661.1 | 2 | 339 | ||
GenBank mRNA | AK002477.1 | - | 2 | 340 | ||
AK018198.1 | - | 2 | 328 | |||
AK028337.1 | - | 1 | 3778 | |||
AK032540.1 | - | 2 | 328 | |||
BC024534.1 | - | 2 | 330 | 4987882 |