Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033136.3 | NP_001028308.1 | 2 | 228 | ||
GenBank mRNA | AK004786.1 | - | 2 | 153 | ||
AK146299.1 | - | 2 | 147 | |||
AK151090.1 | - | 2 | 149 | |||
AK153322.1 | - | 2 | 149 | |||
AK153369.1 | - | 2 | 148 | |||
AK165302.1 | - | 1 | 492 | |||
AK171770.1 | - | 2 | 229 | |||
BC055754.1 | - | 2 | 122 | |||
BC061186.1 | - | 2 | 173 |