Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083317.1 | NP_001076786.1 | 4 | 1291 | ||
NM_026404.2 | NP_080680.2 | 3 | 1399 | |||
GenBank mRNA | AK011633.1 | - | 3 | 1023 | ||
AK018546.1 | - | 3 | 1023 | |||
AK031132.1 | - | 3 | 1023 | |||
AK139953.1 | - | 4 | 859 | |||
AK145812.1 | - | 3 | 1023 | |||
AK164797.1 | - | 4 | 915 | |||
AK170045.1 | - | 4 | 928 | |||
AK182375.1 | - | 1 | 156 | |||
BC006050.1 | - | 2 | 1000 | 3590779 | ||
BC117795.1 | - | 2 | 716 | 40092185 | ||
BC117796.1 | - | 2 | 716 | 40092186 |