Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026406.3 | NP_080682.3 | 9 | 1005 | ||
GenBank mRNA | AK008329.1 | - | 9 | 859 | ||
AK011584.1 | - | 9 | 935 | |||
AK042216.1 | - | 10 | 1000 | |||
AK131966.1 | - | 9 | 925 | |||
AK159772.1 | - | 9 | 898 | |||
BC023113.1 | - | 9 | 952 | 4015975 |