Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026551.3 | NP_080827.2 | 3 | 438 | ||
GenBank mRNA | AK051022.1 | - | 3 | 439 | ||
AK078460.1 | - | 3 | 371 | |||
AK162069.1 | - | 3 | 398 | |||
AK170762.1 | - | 3 | 389 | |||
AK190020.1 | - | 1 | 135 | |||
BC005607.1 | - | 3 | 404 | 3498151 | ||
BC023208.1 | - | 2 | 352 | |||
BC031855.1 | - | 3 | 387 | |||
BC042685.1 | - | 2 | 352 | |||
BC045193.1 | - | 3 | 392 | 5368157 | ||
BC047340.1 | - | 2 | 352 |