Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190385.1 | NP_001177314.1 | 4 | 434 | ||
NM_001190386.1 | NP_001177315.1 | 5 | 714 | |||
NM_026769.5 | NP_081045.2 | 4 | 434 | |||
GenBank mRNA | AK002979.1 | - | 4 | 395 | ||
AK003400.2 | - | 4 | 436 | |||
BC049663.1 | - | 5 | 681 | 6532801 |