Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178111.3 | NP_835212.1 | 4 | 863 | ||
GenBank mRNA | AJ297793.1 | - | 4 | 795 | ||
AK146589.1 | - | 4 | 868 | |||
AK159265.1 | - | 2 | 439 | |||
AK170132.1 | - | 4 | 901 | |||
BC036958.1 | - | 3 | 670 | 4504446 | ||
BC043086.1 | - | 4 | 863 | 6417598 |