Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160214.1 | NP_001153686.1 | 6 | 1915 | ||
NM_211355.2 | NP_997594.1 | 6 | 1915 | |||
GenBank mRNA | AK051239.1 | - | 6 | 1840 | ||
AK152480.1 | - | 6 | 1912 | |||
AK156272.1 | - | 6 | 1877 | |||
AK173331.1 | - | 6 | 1818 | |||
BC064465.1 | - | 6 | 1613 | 6825400 |