Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026891.2 | NP_081167.2 | 22, 23 | 2986 | ||
GenBank mRNA | AK034144.1 | - | 7, 8 | 679 | ||
AK157271.1 | - | 22, 23 | 2987 | |||
BC117555.1 | - | 20, 21 | 2271 | 40098039 |