Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027036.3 | NP_081312.2 | 1 | 598 | ||
GenBank mRNA | AK005588.1 | - | 1 | 597 | ||
BC048744.1 | - | 2 | 558 | |||
BC061030.1 | - | 1 | 594 | 6775221 | ||
BC087910.1 | - | 2 | 484 | |||
BC087934.1 | - | 2 | 554 | |||
CA465179.1 | - | 1 | 597 |