Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027111.3 | NP_081387.1 | 5 | 434 | ||
GenBank mRNA | AK009014.1 | - | 5 | 435 | ||
BC119394.1 | - | 5 | 427 | 8734143 | ||
BC119396.1 | - | 5 | 427 | 8734145 |