Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027111.3 | NP_081387.1 | 5, 6 | 540 | ||
GenBank mRNA | AK009014.1 | - | 5, 6 | 541 | ||
BC119394.1 | - | 5, 6 | 533 | 8734143 | ||
BC119396.1 | - | 5, 6 | 533 | 8734145 |