Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033150.1 | NP_001028322.1 | 4 | 546 | ||
GenBank mRNA | AK122386.1 | - | 630 | |||
AK156652.1 | - | 1 | 168 | |||
AK185623.1 | - | 2 | 136 | |||
AK219223.1 | - | 3 | 211 | |||
BC058984.1 | - | 2 | 137 | |||
CX240883.1 | - | 4 | 546 |