Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243584.1 | NP_001230513.1 | 4 | 485 | ||
NM_001243586.1 | NP_001230515.1 | 3 | 769 | |||
NM_001243587.1 | NP_001230516.1 | 4 | 485 | |||
NM_027162.4 | NP_081438.4 | 3 | 769 | |||
GenBank mRNA | AK003708.1 | - | 546 | |||
AK010175.1 | - | 4 | 439 | |||
AK150749.1 | - | 4 | 439 | |||
BC019515.1 | - | 4 | 396 | 4460171 | ||
BC026740.1 | - | 4 | 445 | 4015105 | ||
BC055812.1 | - | 3 | 769 | 4934728 | ||
CX210298.1 | - | 4 | 469 |