Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027206.2 | NP_081482.1 | 2 | 114 | ||
GenBank mRNA | AF548004.2 | - | 1 | 22 | ||
AK007540.1 | - | 2 | 115 | |||
BC055879.1 | - | 2 | 89 | 3669700 | ||
BC094616.1 | - | 2 | 118 | 573953 |