Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027210.3 | NP_081486.1 | 3 | 818 | ||
NM_028171.2 | NP_082447.1 | 3 | 818 | |||
GenBank mRNA | AK005430.1 | - | 3 | 809 | ||
AK005537.1 | - | 3 | 819 | |||
AK146045.1 | - | 3 | 819 | |||
BC099430.1 | - | 3 | 805 | 30787167 |