Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053085.2 | NP_444315.1 | 3 | 651 | ||
GenBank mRNA | AF142405.1 | - | 3 | 651 | ||
AK030277.1 | - | 3 | 667 | |||
AK030297.1 | - | 3 | 657 | |||
BC119346.1 | - | 3 | 622 | 8734095 | ||
BC120609.1 | - | 3 | 622 | 40129532 |