Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197993.2 | NP_932110.1 | 9 | 2640 | ||
GenBank mRNA | AK012330.1 | - | 1 | 334 | ||
AK177989.1 | - | 1 | 95 | |||
BC023900.1 | - | 9 | 2617 | 5321265 | ||
BC025499.1 | - | 9 | 2664 | 5320398 |