Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026392.1 | NP_080668.1 | 3 | 549 | ||
NM_027415.2 | NP_081691.2 | 3 | 546 | |||
GenBank mRNA | AK003839.1 | - | 3 | 531 | ||
AK005075.1 | - | 3 | 531 | |||
AK008955.1 | - | 3 | 515 | |||
AK151280.1 | - | 3 | 521 | |||
AK167069.1 | - | 3 | 472 | |||
AK169031.1 | - | 3 | 479 | |||
BC011320.1 | - | 3 | 475 | 3979025 |