Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081205.1 | NP_001074674.1 | 2, 3 | 408 | ||
GenBank mRNA | AK014427.1 | - | 2, 3 | 361 | ||
AK075654.1 | - | 2, 3 | 409 |