Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027526.1 | NP_081802.1 | 10 | 1287 | ||
GenBank mRNA | AK018120.1 | - | 9 | 1118 | ||
AK158756.1 | - | 11 | 1313 | |||
BC138284.1 | - | 10 | 1253 | 8861303 | ||
BC145232.1 | - | 10 | 1280 | 9053784 | ||
BC145752.1 | - | 10 | 1253 | 40131055 |