Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195496.1 | NP_001182425.1 | 7 | 1835 | ||
NM_001195497.1 | NP_001182426.1 | 6 | 1787 | |||
NM_001195498.1 | NP_001182427.1 | 7 | 1838 | |||
NM_001195499.1 | NP_001182428.1 | 7 | 1835 | |||
NM_001195500.1 | NP_001182429.1 | 6 | 1787 | |||
NM_027539.5 | NP_081815.3 | 6 | 1790 | |||
GenBank mRNA | AB198721.1 | - | 7 | 1192 | ||
AK049126.1 | - | 6 | 1277 | |||
AK049179.1 | - | 6 | 1239 | |||
AK082633.1 | - | 6 | 1286 | |||
AY968049.1 | - | 7 | 1123 | |||
AY968050.1 | - | 7 | 1126 | |||
AY968051.1 | - | 7 | 1123 | |||
BC056921.1 | - | 6 | 1758 | 6822681 | ||
BC060192.1 | - | 6 | 1091 |