Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027541.4 | NP_081817.2 | 16 | 2414 | ||
GenBank mRNA | AK014398.1 | - | 16 | 2022 | ||
AK088194.1 | - | 16 | 2415 | |||
AK139852.1 | - | 16 | 2047 | |||
AK167435.1 | - | 16 | 2051 | |||
AK185640.1 | - | 11 | ||||
BC006782.1 | - | 16 | 1999 | 3589226 | ||
BC026607.1 | - | 13 | 2226 | |||
BC040787.1 | - | 13 | 2226 |