Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251881.1 | NP_001238810.1 | 5 | 500 | ||
NM_027544.2 | NP_081820.2 | 4 | 929 | |||
GenBank mRNA | AB250379.1 | - | 4 | 812 | ||
AK077129.1 | - | 4 | 812 | |||
BC049764.1 | - | 4 | 790 | 6774330 | ||
CF585840.1 | - | 5 | 512 | |||
CO041745.1 | - | 3 | 610 |