Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027582.3 | NP_081858.2 | 9 | 1110 | ||
GenBank mRNA | AK014937.1 | - | 8 | 989 | ||
AK135492.1 | - | 9 | 1113 | |||
BC051128.1 | - | 9 | 1110 | 6707386 |