Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027733.5 | NP_082009.3 | 1 | 126 | ||
NM_029485.2 | NP_083761.1 | 1 | 126 | |||
GenBank mRNA | AK012241.1 | - | 1 | 98 | ||
AK016341.1 | - | 1 | 98 | |||
AK017218.1 | - | 1 | 102 | |||
AY730532.1 | - | 1 | 126 | |||
BC048524.1 | - | 1 | 75 | |||
BC060950.1 | - | 1 | 79 | 6773733 | ||
BU961109.1 | - | 1 | 129 |