Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027799.2 | NP_082075.2 | 3 | 797 | ||
NM_146024.1 | NP_666136.1 | 3 | 797 | |||
GenBank mRNA | AK017451.1 | - | 3 | 526 | ||
BC023133.1 | - | 3 | 649 | 3592723 |