Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081321.1 | NP_001074790.1 | 27 | 3334 | ||
GenBank mRNA | AK018509.1 | - | 3 | 1260 | ||
AK136537.1 | - | 3 | 2404 | |||
AK172992.1 | - | 26 | 5031 | |||
BC021408.1 | - | 14 | 1717 |