Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294278.1 | NP_001281207.1 | 4 | 995 | ||
NM_027837.3 | NP_082113.2 | 4 | 1036 | |||
GenBank mRNA | AB219124.1 | - | 4 | 569 | ||
AK018615.1 | - | 4 | 1037 | |||
BC128289.1 | - | 4 | 629 | 40102442 | ||
BC128290.1 | - | 4 | 670 | 40102443 |