Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027875.1 | NP_082151.1 | 2 | 396 | ||
GenBank mRNA | AK004654.1 | - | 2 | 378 |