Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027895.1 | NP_082171.1 | 4 | 563 | ||
GenBank mRNA | AK004783.1 | - | 4 | 547 | ||
AK032030.1 | - | 2 | 184 | |||
AK154642.1 | - | 4 | 476 | |||
AK205345.1 | - | 1 | 12 | |||
BC037093.1 | - | 4 | 584 | |||
BC151153.1 | - | 4 | 489 | 9088047 |