Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035854.2 | NP_001030931.1 | 11 | 1506 | ||
NM_027915.3 | NP_082191.1 | 11 | 1506 | |||
GenBank mRNA | AK004975.1 | - | 11 | 1430 | ||
AK030505.1 | - | 11 | 1500 | |||
AK032617.1 | - | 11 | 1469 | |||
AK138434.1 | - | 11 | 1507 | |||
BC013699.1 | - | 7 | 892 | |||
BC046772.1 | - | 11 | 1478 | 5708062 |