Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146012.2 | NP_001139484.2 | 4 | 496 | ||
NM_001301651.1 | NP_001288580.1 | 4 | 448 | |||
NM_001301652.1 | NP_001288581.1 | 3 | 729 | |||
GenBank mRNA | AK006177.1 | - | 4 | 448 | ||
AK032854.1 | - | 4 | 466 | |||
AK156843.1 | - | 4 | 466 | |||
BC052536.1 | - | 3 | 645 | |||
BC059860.1 | - | 3 | 729 |