Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027994.1 | NP_082270.1 | 10 | 2090 | ||
GenBank mRNA | AK129225.1 | - | 10 | 2051 | ||
AK144869.1 | - | 10 | 2082 | |||
AK158080.1 | - | 10 | 2086 | |||
BC057457.1 | - | 3 | 733 |