Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027994.1 | NP_082270.1 | 15 | 3969 | ||
GenBank mRNA | AK009683.1 | - | 4 | 608 | ||
AK078117.1 | - | 6 | 1196 | |||
AK129225.1 | - | 15 | 3930 | |||
BC027142.1 | - | 3 | 336 | |||
BC057457.1 | - | 8 | 2612 |