Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027999.1 | NP_082275.1 | 15 | 1383 | ||
GenBank mRNA | AK009476.1 | - | 15 | 1363 | ||
AK129228.1 | - | 11 | 2651 | |||
AK153920.1 | - | 15 | 1365 | |||
BC023723.1 | - | 15 | 1333 | |||
BC089002.1 | - | 15 | 1352 | |||
BC118934.1 | - | 15 | 1358 | 40090953 | ||
BC118935.1 | - | 15 | 1358 | 40090954 |