Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028006.2 | NP_082282.1 | 7 | 592 | ||
GenBank mRNA | AK010005.1 | - | 7 | 586 | ||
AK040492.1 | - | 1 | 1316 | |||
AK041416.1 | - | 7 | 557 | |||
AK041986.1 | - | 174 | ||||
AK087403.1 | - | 7 | 547 | |||
AK148039.1 | - | 7 | 592 | |||
BC038654.1 | - | 4 | 547 | |||
BC062179.1 | - | 7 | 589 | 6333817 |