Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028071.3 | NP_082347.1 | 4 | 593 | ||
GenBank mRNA | AK007994.1 | - | 4 | 555 | ||
AK008085.1 | - | 4 | 555 | |||
AK008597.1 | - | 1 | 20 | |||
AK010052.1 | - | 4 | 555 | |||
AK039644.1 | - | 4 | 555 | |||
AK148854.1 | - | 4 | 559 | |||
AK150888.1 | - | 4 | 555 | |||
AK151690.1 | - | 4 | 555 | |||
AK157694.1 | - | 4 | 555 | |||
AK172403.1 | - | 4 | 555 | |||
AK210405.1 | - | 2 | 160 | |||
AK213320.1 | - | 2 | 160 | |||
BC010249.1 | - | 4 | 548 | 3155195 | ||
BC011068.1 | - | 4 | 548 | 3157387 | ||
CT010261.1 | - | 4 | 414 |