Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133928.2 | NP_598689.1 | 3 | 320 | ||
GenBank mRNA | AK010479.1 | - | 3 | 291 | ||
AK012745.1 | - | 3 | 429 | |||
BC019405.1 | - | 3 | 320 | 5066665 |