Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162996.1 | NP_001156468.1 | 7, 8 | 630 | ||
NM_028153.1 | NP_082429.1 | 7, 8 | 687 | |||
GenBank mRNA | AK005562.1 | - | 7, 8 | 653 | ||
AK184916.1 | - | 3, 4 | 243 | |||
AK213701.1 | - | 2, 3 | 144 | |||
BC055476.1 | - | 7, 8 | 630 | 4218133 |