Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028222.1 | NP_082498.1 | 7 | 599 | ||
GenBank mRNA | AK146312.1 | - | 7 | 515 | ||
BC049694.1 | - | 7 | 583 | 6705916 |