Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028360.2 | NP_082636.3 | 9 | 937 | ||
NM_029704.2 | NP_083980.2 | 8 | 898 | |||
GenBank mRNA | AK013368.1 | - | 9 | 890 | ||
AK034075.1 | - | 9 | 927 | |||
AK177481.1 | - | 1 | 48 | |||
AK185142.1 | - | 1 | 48 | |||
BC026866.1 | - | 7 | 581 | |||
BC037111.1 | - | 8 | 661 | 5370464 |