Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028532.3 | NP_082808.1 | 5, 6 | 454 | ||
NM_199455.2 | NP_955534.1 | 6, 7 | 679 | |||
GenBank mRNA | AK006802.1 | - | 5, 6 | 441 | ||
BC087870.1 | - | 6, 7 | 675 | 6707505 | ||
BC104382.1 | - | 5, 6 | 453 | |||
BC104383.1 | - | 5, 6 | 454 | |||
BY706785.1 | - | 5, 6 | 439 |