Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267872.1 | NP_001254801.1 | 6 | 3220 | ||
NM_001285936.1 | NP_001272865.1 | 7 | 3354 | |||
NM_001285937.1 | NP_001272866.1 | 6 | 3297 | |||
NM_028673.1 | NP_082949.1 | 7 | 3270 | |||
GenBank mRNA | AB777270.1 | - | 7 | 3354 |