Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syvn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164709.1 | NP_001158181.1 | 4 | 472 | ||
NM_028769.5 | NP_083045.4 | 4 | 839 | |||
GenBank mRNA | AK004688.2 | - | 4 | 326 | ||
AK122558.1 | - | 3 | 441 | |||
AK143720.1 | - | 4 | 351 | |||
AK172097.1 | - | 4 | 351 | |||
BC042199.1 | - | 4 | 334 | 4953165 | ||
BC046829.1 | - | 4 | 472 | 6811574 | ||
BC057917.1 | - | 4 | 753 | 5136398 | ||
BC080722.1 | - | 4 | 357 | 5323695 |