Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145827.1 | NP_001139299.1 | 7 | 974 | ||
NM_028800.3 | NP_083076.3 | 5 | 620 | |||
GenBank mRNA | AB102946.1 | - | 5 | 451 | ||
AK154780.1 | - | 7 | 965 | |||
AK192521.1 | - | 1 | 64 | |||
AY336057.1 | - | 5 | 605 | |||
BC046981.1 | - | 5 | 579 | 6478265 |