Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023741.2 | NP_076230.1 | 6 | 721 | ||
GenBank mRNA | AF230921.1 | - | 6 | 707 | ||
AF230922.1 | - | 6 | 710 | |||
AF230923.1 | - | 7 | 923 | |||
AF466150.1 | - | 6 | 722 | |||
AK005568.1 | - | 6 | 727 | |||
AK167369.1 | - | 6 | 728 | |||
BC100326.1 | - | 6 | 715 | 30787396 |