Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165935.1 | NP_001159407.1 | 2 | 151 | ||
NM_028844.3 | NP_083120.2 | 3 | 481 | |||
GenBank mRNA | AK006808.1 | - | 3 | 482 | ||
AK046479.1 | - | 407 | ||||
BC100725.1 | - | 4 | 344 | 40045263 | ||
BC100735.1 | - | 4 | 358 | |||
BX528932.1 | - | 11 | ||||
DV646415.1 | - | 2 | 153 |